Merck
  • Simultaneous determination of pyrethroids and pyrethrins by dispersive liquid-liquid microextraction and liquid chromatography triple quadrupole mass spectrometry in environmental samples.
Simultaneous determination of pyrethroids and pyrethrins by dispersive liquid-liquid microextraction and liquid chromatography triple quadrupole mass spectrometry in environmental samples.
Analytical and bioanalytical chemistry (2017-06-11)
Alexander Ccanccapa-Cartagena, Ana Masiá, Yolanda Picó
ABSTRACT
A simple and environmentally friendly dispersive liquid-liquid microextraction (DLLME) method coupled with electrospray ionization liquid chromatography triple quadrupole mass spectrometry (LC-QqQ-MS/MS) was developed for the simultaneous determination of 17 synthetic and natural pyrethroids. A comparison of solid-phase extraction (SPE) versus DLLME for water samples and only "dilute and shoot" versus the additional extract cleanup by DLLME for sediment samples is reported. Chloroform was the extracting solvent in the DLLME technique for both water and sediment samples. Ultrasonic energy was applied to fully extract the analytes into fine droplets, providing high recoveries in short times. Method detection limits (MDLs) ranged from 0.12 to 0.62 ng L
Product Description
Supelco
Tefluthrin, PESTANAL®, analytical standard
Supelco
Acrinathrin, PESTANAL®, analytical standard
Supelco
Etofenprox, PESTANAL®, analytical standard
Supelco
Esfenvalerate, PESTANAL®, analytical standard
Supelco
Flumethrin, PESTANAL®, analytical standard