- In vitro inhibition of UGT1A3, UGT1A4 by ursolic acid and oleanolic acid and drug-drug interaction risk prediction.
In vitro inhibition of UGT1A3, UGT1A4 by ursolic acid and oleanolic acid and drug-drug interaction risk prediction.
Xenobiotica; the fate of foreign compounds in biological systems (2016-09-08)
Hongbo Xie, Jie Wu, Dan Liu, Mingyi Liu, Hong Zhang, Shibo Huang, Yuqing Xiong, Chunhua Xia
PMID27600106
ABSTRACT
1. Ursolic acid (UA) and oleanolic acid (OA) may have important activity relevant to health and disease prevention. Thus, we studied the activity of UA and OA on UDP-glucuronosyltransferases (UGTs) and used trifluoperazine as a probe substrate to test UGT1A4 activity. Recombinant UGT-catalyzed 4-methylumbelliferone (4-MU) glucuronidation was used as a probe reaction for other UGT isoforms. 2. UA and OA inhibited UGT1A3 and UGT1A4 activity but did not inhibit other tested UGT isoforms. 3. UA-mediated inhibition of UGT1A3 catalyzed 4-MU-β-d-glucuronidation was via competitive inhibition (IC