Merck
  • Isomerization versus dissociation of phenylalanylglycyltryptophan radical cations.
Isomerization versus dissociation of phenylalanylglycyltryptophan radical cations.
Physical chemistry chemical physics : PCCP (2017-06-21)
Xiaoyan Mu, Justin Kai-Chi Lau, Cheuk-Kuen Lai, K W Michael Siu, Alan C Hopkinson, Ivan K Chu
ABSTRACT
Four isomers of the radical cation of tripeptide phenylalanylglycyltryptophan, in which the initial location of the radical center is well defined, have been isolated and their collision-induced dissociation (CID) spectra examined. These ions, the π-centered [FGW
Sigma-Aldrich
Fmoc-Phe(4-I)-OH, ≥97.0%