Merck
  • Comprehensive evaluation of the bactericidal activities of free bile acids in the large intestine of humans and rodents.
Comprehensive evaluation of the bactericidal activities of free bile acids in the large intestine of humans and rodents.
Journal of lipid research (2017-04-14)
Masamichi Watanabe, Satoru Fukiya, Atsushi Yokota
ABSTRACT
In addition to functioning as detergents that aid digestion of dietary lipids in the intestine, some bile acids have been shown to exhibit antimicrobial activity. However, detailed information on the bactericidal activities of the diverse molecular species of bile acid in humans and rodents is largely unknown. Here, we investigated the toxicity of 14 typical human and rodent free bile acids (FBAs) by monitoring intracellular pH, membrane integrity, and viability of a human intestinal bacterium, Bifidobacterium breve Japan Collection of Microorganisms (JCM) 1192T, upon exposure to these FBAs. Of all FBAs evaluated, deoxycholic acid (DCA) and chenodeoxycholic acid displayed the highest toxicities. Nine FBAs common to humans and rodents demonstrated that α-hydroxy-type bile acids are more toxic than their oxo-derivatives and β-hydroxy-type epimers. In five rodent-specific FBAs, β-muricholic acid and hyodeoxycholic acid showed comparable toxicities at a level close to DCA. Similar trends were observed for the membrane-damaging effects and bactericidal activities to Blautia coccoides JCM 1395T and Bacteroides thetaiotaomicron DSM 2079T, commonly represented in the human and rodent gut microbiota. These findings will help us to determine the fundamental properties of FBAs and better understand the role of FBAs in the regulation of gut microbiota composition.
Sigma-Aldrich
5(6)-Carboxyfluorescein diacetate, BioReagent, suitable for fluorescence, ≥90.0% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
β-Muricholic Acid, ≥98% (HPLC)
Pricing and availability is not currently available.