Merck
  • Bub1 positions Mad1 close to KNL1 MELT repeats to promote checkpoint signalling.
Bub1 positions Mad1 close to KNL1 MELT repeats to promote checkpoint signalling.
Nature communications (2017-06-13)
Gang Zhang, Thomas Kruse, Blanca López-Méndez, Kathrine Beck Sylvestersen, Dimitriya H Garvanska, Simone Schopper, Michael Lund Nielsen, Jakob Nilsson
ABSTRACT
Proper segregation of chromosomes depends on a functional spindle assembly checkpoint (SAC) and requires kinetochore localization of the Bub1 and Mad1/Mad2 checkpoint proteins. Several aspects of Mad1/Mad2 kinetochore recruitment in human cells are unclear and in particular the underlying direct interactions. Here we show that conserved domain 1 (CD1) in human Bub1 binds directly to Mad1 and a phosphorylation site exists in CD1 that stimulates Mad1 binding and SAC signalling. Importantly, fusion of minimal kinetochore-targeting Bub1 fragments to Mad1 bypasses the need for CD1, revealing that the main function of Bub1 is to position Mad1 close to KNL1 MELT repeats. Furthermore, we identify residues in Mad1 that are critical for Mad1 functionality, but not Bub1 binding, arguing for a direct role of Mad1 in the checkpoint. This work dissects functionally relevant molecular interactions required for spindle assembly checkpoint signalling at kinetochores in human cells.
Sigma-Aldrich
Streptavidin−FITC from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥5 units/mg protein
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Cdc20 Antibody, clone AR12, clone AR12, Chemicon®, from mouse
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder