Merck
  • Long-term, hormone-responsive organoid cultures of human endometrium in a chemically defined medium.
Long-term, hormone-responsive organoid cultures of human endometrium in a chemically defined medium.
Nature cell biology (2017-04-11)
Margherita Y Turco, Lucy Gardner, Jasmine Hughes, Tereza Cindrova-Davies, Maria J Gomez, Lydia Farrell, Michael Hollinshead, Steven G E Marsh, Jan J Brosens, Hilary O Critchley, Benjamin D Simons, Myriam Hemberger, Bon-Kyoung Koo, Ashley Moffett, Graham J Burton
ABSTRACT
In humans, the endometrium, the uterine mucosal lining, undergoes dynamic changes throughout the menstrual cycle and pregnancy. Despite the importance of the endometrium as the site of implantation and nutritional support for the conceptus, there are no long-term culture systems that recapitulate endometrial function in vitro. We adapted conditions used to establish human adult stem-cell-derived organoid cultures to generate three-dimensional cultures of normal and decidualized human endometrium. These organoids expand long-term, are genetically stable and differentiate following treatment with reproductive hormones. Single cells from both endometrium and decidua can generate a fully functional organoid. Transcript analysis confirmed great similarity between organoids and the primary tissue of origin. On exposure to pregnancy signals, endometrial organoids develop characteristics of early pregnancy. We also derived organoids from malignant endometrium, and so provide a foundation to study common diseases, such as endometriosis and endometrial cancer, as well as the physiology of early gestation.
Sigma-Aldrich
Dispase® II, protease
Sigma-Aldrich
IgG from human serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Proteinase K from Tritirachium album, buffered aqueous glycerol solution, ≥800 units/mL
Sigma-Aldrich
SIGMAFAST 3,3′-Diaminobenzidine tablets, tablet, To prepare 1 mL
Sigma-Aldrich
p-Xylene-bis(N-pyridinium bromide), ≥95% (TLC)
Sigma-Aldrich
Progesterone-Water Soluble, powder, BioReagent, suitable for cell culture
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
Collagenase from Clostridium histolyticum, Type V, ≥1 FALGPA units/mg solid, >125 CDU/mg solid
Sigma-Aldrich
Ribonuclease A from bovine pancreas, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Penicillin-Streptomycin, with 10,000 units penicillin and 10 mg streptomycin per mL in 0.9% NaCl, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
β-Estradiol-Water Soluble, BioReagent, suitable for cell culture
Sigma-Aldrich
Formalin, 10%, Neutral Buffered, Wintergreen Scented, histological fixative, pH 6.9-7.1
Sigma-Aldrich
Y-27632-CAS 331752-47-7-Calbiochem, Y-27632A, CAS 331752-47-7, is a cell-permeable, reversible, inhibitor of Rho kinases (Ki = 140 nM for p160ROCK). Enhances survival & cloning efficiency of ESC without affecting their pluripotency.