Merck
  • Highly Viscous Binary Solvents: DMSO-d6/Glycerol and DMSO-d6/Glycerol-d8 for Polar and Apolar Mixture Analysis by NMR.
Highly Viscous Binary Solvents: DMSO-d6/Glycerol and DMSO-d6/Glycerol-d8 for Polar and Apolar Mixture Analysis by NMR.
AZo Journal of Materials Online (2016)
Lameiras P and Nuzillard J-M.