Merck
  • Elucidation of Surface Species through in Situ FTIR Spectroscopy of Carbon Dioxide Adsorption on Amine-Grafted SBA-15.
Elucidation of Surface Species through in Situ FTIR Spectroscopy of Carbon Dioxide Adsorption on Amine-Grafted SBA-15.
ChemSusChem (2016-08-31)
Guo Shiou Foo, Jason J Lee, Chia-Hsin Chen, Sophia E Hayes, Carsten Sievers, Christopher W Jones
ABSTRACT
The nature of the surface species formed through the adsorption of CO
Sigma-Aldrich
(N,N-Dimethylaminopropyl)trimethoxysilane, 96%