- Integration of Organic Electrochemical and Field-Effect Transistors for Ultraflexible, High Temporal Resolution Electrophysiology Arrays.
Integration of Organic Electrochemical and Field-Effect Transistors for Ultraflexible, High Temporal Resolution Electrophysiology Arrays.
Advanced materials (Deerfield Beach, Fla.) (2016-10-08)
Wonryung Lee, Dongmin Kim, Jonathan Rivnay, Naoji Matsuhisa, Thomas Lonjaret, Tomoyuki Yokota, Hiromu Yawo, Masaki Sekino, George G Malliaras, Takao Someya
PMID27717052
ABSTRACT
Integration of organic electrochemical transistors and organic field-effect transistors is successfully realized on a 600 nm thick parylene film toward an electrophysiology array. A single cell of an integrated device and a 2 × 2 electrophysiology array succeed in detecting electromyogram with local stimulation of the motor nerve bundle of a transgenic rat by a laser pulse.