Merck
  • Partial bladder outlet obstruction is associated with decreased expression and function of the small-conductance Ca
Partial bladder outlet obstruction is associated with decreased expression and function of the small-conductance Ca
International urology and nephrology (2016-11-16)
Ning Li, Xiaoning He, Zizheng Li, Yili Liu, Ping Wang
ABSTRACT
Partial bladder outlet obstruction (PBOO) usually induces overactive bladder (OAB) associated with detrusor overactivity (DO) which is related to the increased contractility of detrusor smooth muscle (DSM). The pharmacological activation of small-conductance Ca Two weeks after surgically inducing PBOO in female guinea pigs, cystometry indicated that DO was achieved. Using this animal model, we conducted quantitative reverse transcription-polymerase chain reaction (qRT-PCR) and isometric tension recordings. The qRT-PCR experiments indicated that PBOO DSM had reduced SK channel mRNA expression. Isometric tension recordings showed a decreased inhibitory effect of NS309 on spontaneous phasic and electrical field stimulation-induced contractions via the activation of SK channels in PBOO DSM. This study presents the novel finding that PBOO is associated with attenuated DSM SK channel expression and function, which results in increased DSM contractility and contributes to DO. Therefore, SK channels could be a therapeutic target to control OAB.
Sigma-Aldrich
TRAM-34, ≥98% (HPLC), solid
Sigma-Aldrich
NS309, ≥98% (HPLC), solid