Merck
  • The granulocytes are the main immunocompetent hemocytes in Crassostrea gigas.
The granulocytes are the main immunocompetent hemocytes in Crassostrea gigas.
Developmental and comparative immunology (2016-10-04)
Weilin Wang, Meijia Li, Lingling Wang, Hao Chen, Zhaoqun Liu, Zhihao Jia, Limei Qiu, Linsheng Song
ABSTRACT
Hemocytes comprise diverse cell types with morphological and functional heterogeneity and play indispensable roles in immunological homeostasis of invertebrates. The morphological classification of different hemocytes in mollusk has been studied since the 1970's, yet the involvement of the different sub-populations in immune functions is far from clear. In the present study, three types of hemocytes were morphologically identified and separated as agranulocytes, semi-granulocytes and granulocytes by flow cytometry and Percoll® density gradient centrifugation. The granulocytes were characterized functionally as the main phagocytic and encapsulating population, while semi-granulocytes and agranulocytes exhibited low or no such capacities, respectively. Meanwhile, the lysosome activity and the productions of ROS and NO were all mainly concentrated in granulocytes under both normal and immune-activated situations. Further, the mRNA transcripts of some immune related genes, including CgTLR, CgClathrin, CgATPeV, CgLysozyme, CgDefensin and CgIL-17, were mainly expressed in granulocytes, lower in semi-granulocytes and agranulocytes. These results collectively suggested that the granulocytes were the main immunocompetent hemocytes in oyster C. gigas, and a differentiation relationship among these three sub-population hemocytes was inferred based on the gradual changes in morphological, functional and molecular features.
Sigma-Aldrich
Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green, aqueous suspension, 1.0 μm mean particle size
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)