Merck
  • Regulation of PIK3C3/VPS34 complexes by MTOR in nutrient stress-induced autophagy.
Regulation of PIK3C3/VPS34 complexes by MTOR in nutrient stress-induced autophagy.
Autophagy (2013-09-10)
Hai-Xin Yuan, Ryan C Russell, Kun-Liang Guan
ABSTRACT
Autophagy is a cellular defense response to stress conditions, such as nutrient starvation. The type III phosphatidylinositol (PtdIns) 3-kinase, whose catalytic subunit is PIK3C3/VPS34, plays a critical role in intracellular membrane trafficking and autophagy induction. PIK3C3 forms multiple complexes and the ATG14-containing PIK3C3 is specifically involved in autophagy induction. Mechanistic target of rapamycin (MTOR) complex 1, MTORC1, is a key cellular nutrient sensor and integrator to stimulate anabolism and inhibit catabolism. Inactivation of TORC1 by nutrient starvation plays a critical role in autophagy induction. In this report we demonstrated that MTORC1 inactivation is critical for the activation of the autophagy-specific (ATG14-containing) PIK3C3 kinase, whereas it has no effect on ATG14-free PIK3C3 complexes. MTORC1 inhibits the PtdIns 3-kinase activity of ATG14-containing PIK3C3 by phosphorylating ATG14, which is required for PIK3C3 inhibition by MTORC1 both in vitro and in vivo. Our data suggest a mechanistic link between amino acid starvation and autophagy induction via the direct activation of the autophagy-specific PIK3C3 kinase.
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
L-α-Phosphatidylinositol sodium salt from Glycine max (soybean), ≥99%