Merck
  • A novel GABRG2 mutation, p.R136*, in a family with GEFS+ and extended phenotypes.
A novel GABRG2 mutation, p.R136*, in a family with GEFS+ and extended phenotypes.
Neurobiology of disease (2014-01-11)
Ann J Johnston, Jing-Qiong Kang, Wangzhen Shen, William O Pickrell, Thomas D Cushion, Jeffrey S Davies, Kristin Baer, Jonathan G L Mullins, Carrie L Hammond, Seo-Kyung Chung, Rhys H Thomas, Cathy White, Phil E M Smith, Robert L Macdonald, Mark I Rees
ABSTRACT
Genetic mutations in voltage-gated and ligand-gated ion channel genes have been identified in a small number of Mendelian families with genetic generalised epilepsies (GGEs). They are commonly associated with febrile seizures (FS), childhood absence epilepsy (CAE) and particularly with generalised or genetic epilepsy with febrile seizures plus (GEFS+). In clinical practice, despite efforts to categorise epilepsy and epilepsy families into syndromic diagnoses, many generalised epilepsies remain unclassified with a presumed genetic basis. During the systematic collection of epilepsy families, we assembled a cohort of families with evidence of GEFS+ and screened for variations in the γ2 subunit of the γ-aminobutyric acid (GABA) type A receptor gene (GABRG2). We detected a novel GABRG2(p.R136*) premature translation termination codon in one index-case from a two-generation nuclear family, presenting with an unclassified GGE, a borderline GEFS+ phenotype with learning difficulties and extended behavioural presentation. The GABRG2(p.R136*) mutation segregates with the febrile seizure component of this family's GGE and is absent in 190 healthy control samples. In vitro expression assays demonstrated that γ2(p.R136*) subunits were produced, but had reduced cell-surface and total expression. When γ2(p.R136*) subunits were co-expressed with α1 and β2 subunits in HEK 293T cells, GABA-evoked currents were reduced. Furthermore, γ2(p.R136*) subunits were highly-expressed in intracellular aggregations surrounding the nucleus and endoplasmic reticulum (ER), suggesting compromised receptor trafficking. A novel GABRG2(p.R136*) mutation extends the spectrum of GABRG2 mutations identified in GEFS+ and GGE phenotypes, causes GABAA receptor dysfunction, and represents a putative epilepsy mechanism.
Sigma-Aldrich
Anti-GABA A Receptor β 2,3 Chain Antibody, clone BD17, clone BD17, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABAA Receptor (γ2 subunit) (extracellular) antibody produced in rabbit, affinity isolated antibody, lyophilized powder
Sigma-Aldrich
Horse Serum, Donor herd, USA origin, sterile-filtered, suitable for cell culture, suitable for hybridoma