Merck
  • Elongation factor 2 kinase promotes cell survival by inhibiting protein synthesis without inducing autophagy.
Elongation factor 2 kinase promotes cell survival by inhibiting protein synthesis without inducing autophagy.
Cellular signalling (2016-01-23)
Claire E J Moore, Xuemin Wang, Jianling Xie, Jo Pickford, John Barron, Sergio Regufe da Mota, Matthias Versele, Christopher G Proud
ABSTRACT
Eukaryotic elongation factor 2 kinase (eEF2K) inhibits the elongation stage of protein synthesis by phosphorylating its only known substrate, eEF2. eEF2K is tightly regulated by nutrient-sensitive signalling pathways. For example, it is inhibited by signalling through mammalian target of rapamycin complex 1 (mTORC1). It is therefore activated under conditions of nutrient deficiency. Here we show that inhibiting eEF2K or knocking down its expression renders cancer cells sensitive to death under nutrient-starved conditions, and that this is rescued by compounds that block protein synthesis. This implies that eEF2K protects nutrient-deprived cells by inhibiting protein synthesis. Cells in which signalling through mTORC1 is highly active are very sensitive to nutrient withdrawal. Inhibiting mTORC1 protects them. Our data reveal that eEF2K makes a substantial contribution to the cytoprotective effect of mTORC1 inhibition. eEF2K is also reported to promote another potentially cytoprotective process, autophagy. We have used several approaches to test whether inhibition or loss of eEF2K affects autophagy under a variety of conditions. We find no evidence that eEF2K is involved in the activation of autophagy in the cell types we have studied. We conclude that eEF2K protects cancer cells against nutrient starvation by inhibiting protein synthesis rather than by activating autophagy.
Sigma-Aldrich
Anti-GAPDH antibody, Mouse monoclonal, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
E-64d, protease inhibitor
Sigma-Aldrich
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)
Sigma-Aldrich
IPTG, ≥99% (TLC), ≤0.1% Dioxane
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GST Antibody, Cytiva 27-4577-01
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP)
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Cycloheximide, ≥95% (HPLC)
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O