Merck
  • A mechanism-based affinity-labeling agent for possible use in isolating N-acetylglucosaminidase.
A mechanism-based affinity-labeling agent for possible use in isolating N-acetylglucosaminidase.
Bioorganic & medicinal chemistry letters (2001-06-27)
M Ichikawa, Y Ichikawa
ABSTRACT
We have prepared several mechanism-based affinity-labeling agents for possible use in isolating N-acetylglucosaminidase, in which an N-acetylglucosamine is linked to an o-monofluoro- or difluoro-methyl phenoxy glycoside with or without a cleavable disulfide group in the tether to biotin.
Sigma-Aldrich
Biotin disulfide N-hydroxysuccinimide ester, >98%, powder
Sigma-Aldrich
Sulfo-NHS-LC-LC-Biotin, ≥95%