Merck
  • The spectral heterogeneity and size distribution of the carbon dots derived from time-resolved fluorescence studies.
The spectral heterogeneity and size distribution of the carbon dots derived from time-resolved fluorescence studies.
Physical chemistry chemical physics : PCCP (2016-11-03)
Ying-Feng Hsu, Yu-Hsun Chen, Chih-Wei Chang
ABSTRACT
We performed comprehensive studies on the photoluminescence properties of the carbon dots prepared from citric acid (C-dots
Sigma-Aldrich
Coumarin 343, Dye content 97 %