Merck
ASK1 is essential for JNK/SAPK activation by TRAF2.
Molecular cell (1998-10-17)
H Nishitoh, M Saitoh, Y Mochida, K Takeda, H Nakano, M Rothe, K Miyazono, H Ichijo
ABSTRACT
Tumor necrosis factor (TNF)-induced activation of the c-jun N-terminal kinase (JNK, also known as SAPK; stress-activated protein kinase) requires TNF receptor-associated factor 2 (TRAF2). The apoptosis signal-regulating kinase 1 (ASK1) is activated by TNF and stimulates JNK activation. Here we show that ASK1 interacts with members of the TRAF family and is activated by TRAF2, TRAF5, and TRAF6 overexpression. A truncated derivative of TRAF2, which inhibits JNK activation by TNF, blocks TNF-induced ASK1 activation. A catalytically inactive mutant of ASK1 is a dominant-negative inhibitor of TNF- and TRAF2-induced JNK activation. In untransfected mammalian cells, ASK1 rapidly associates with TRAF2 in a TNF-dependent manner. Thus, ASK1 is a mediator of TRAF2-induced JNK activation.