Merck
  • Rabex-5 is a lenalidomide target molecule that negatively regulates TLR-induced type 1 IFN production.
Rabex-5 is a lenalidomide target molecule that negatively regulates TLR-induced type 1 IFN production.
Proceedings of the National Academy of Sciences of the United States of America (2016-09-08)
David Millrine, Mami Tei, Yohannes Gemechu, Tadamitsu Kishimoto
ABSTRACT
Immunomodulatory drugs (IMiDs) are a family of compounds derived from thalidomide. Binding of the IMiD molecule to the Lon protease Cereblon initiates the degradation of substrates via the ubiquitin proteasome pathway. Here, we show that Cereblon forms a complex with Rabex-5, a regulator of immune homeostasis. Treatment with lenalidomide prevented the association of Cereblon with Rabex-5. Conversely, mutation of the IMiD binding site increased Cereblon-Rabex-5 coimmunoprecipitation. The thalidomide binding region of Cereblon therefore regulates the formation of this complex. Knockdown of Rabex-5 in the THP-1 macrophage cell line up-regulated Toll-like receptor (TLR)-induced cytokine and type 1 IFN production via a STAT1/IRF activating pathway. Thus, we identify Rabex-5 as a IMiD target molecule that functions to restrain TLR activated auto-immune promoting pathways. We propose that release of Rabex-5 from complex with Cereblon enables the suppression of immune responses, contributing to the antiinflammatory properties of IMiDs.
Sigma-Aldrich
Anti-Rabex-5 (N-terminal) antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2-Peroxidase (HRP) antibody produced in mouse, clone M2, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
MISSION® Lentiviral Packaging Mix, Production of replication incompetent viral particles
Sigma-Aldrich
Lipopolysaccharides from Salmonella enterica serotype minnesota, purified by phenol extraction
Millipore
ANTI-FLAG® M2 Affinity Gel, purified immunoglobulin, buffered aqueous glycerol solution