Merck
  • Kinesin-1 sorting in axons controls the differential retraction of arbor terminals.
Kinesin-1 sorting in axons controls the differential retraction of arbor terminals.
Journal of cell science (2016-08-10)
Takeshi Seno, Tatsuki Ikeno, Kousuke Mennya, Masayuki Kurishita, Narumi Sakae, Makoto Sato, Hiroki Takada, Yoshiyuki Konishi
ABSTRACT
The ability of neurons to generate multiple arbor terminals from a single axon is crucial for establishing proper neuronal wiring. Although growth and retraction of arbor terminals are differentially regulated within the axon, the mechanisms by which neurons locally control their structure remain largely unknown. In the present study, we found that the kinesin-1 (Kif5 proteins) head domain (K5H) preferentially marks a subset of arbor terminals. Time-lapse imaging clarified that these arbor terminals were more stable than others, because of a low retraction rate. Local inhibition of kinesin-1 in the arbor terminal by chromophore-assisted light inactivation (CALI) enhanced the retraction rate. The microtubule turnover was locally regulated depending on the length from the branching point to the terminal end, but did not directly correlate with the presence of K5H. By contrast, F-actin signal values in arbor terminals correlated spatiotemporally with K5H, and inhibition of actin turnover prevented retraction. Results from the present study reveal a new system mediated by kinesin-1 sorting in axons that differentially controls stability of arbor terminals.
Sigma-Aldrich
Monoclonal Anti-Tubulin, Tyrosine antibody produced in mouse, clone TUB-1A2, ascites fluid
Sigma-Aldrich
MISSION® esiRNA, targeting human TTL
Sigma-Aldrich
Anti-Kinesin Antibody, heavy chain, a.a.420-445, clone H2, clone H2, Chemicon®, from mouse
Sigma-Aldrich
Anti-β-Tubulin−Cy3 antibody, Mouse monoclonal, clone TUB 2.1, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-Tubulin, Acetylated antibody produced in mouse, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Anti-Tubulin Antibody, Detyrosinated, Chemicon®, from rabbit