Merck
Synthesis of bicyclo[5.3.0]azulene derivatives.
Nature protocols (2009-07-21)
Donald D Nolting, Michael Nickels, Ronald Price, John C Gore, Wellington Pham
ABSTRACT
Azulene has been recognized for its application in medicinal therapy against inflammation. Recently, azulene analogs have been used in optical technology. Nevertheless, synthesis of this family of compounds is always associated with multiple challenges. In this protocol, we describe a time-efficient and cost-effective procedure for the preparation of azulene derivatives from 2-hydroxycyclohepta-2,4,6-trienone (tropolone), a readily available starting material. The technique illustrated here involves a cycloaddition reaction of a lactone with the in situ-generated vinyl ether from 2,2-dimethoxypropane during the thermolysis reaction. The three-step synthesis should take <4 d, resulting in an overall yield of 74% with a final step yield of 91%.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Tropolone, 98%
Sigma-Aldrich
Dichloromethane, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Silicone oil, high temperature