Merck
  • BK channels in microglia are required for morphine-induced hyperalgesia.
BK channels in microglia are required for morphine-induced hyperalgesia.
Nature communications (2016-06-01)
Yoshinori Hayashi, Saori Morinaga, Jing Zhang, Yasushi Satoh, Andrea L Meredith, Takahiro Nakata, Zhou Wu, Shinichi Kohsaka, Kazuhide Inoue, Hiroshi Nakanishi
ABSTRACT
Although morphine is a gold standard medication, long-term opioid use is associated with serious side effects, such as morphine-induced hyperalgesia (MIH) and anti-nociceptive tolerance. Microglia-to-neuron signalling is critically involved in pain hypersensitivity. However, molecules that control microglial cellular state under chronic morphine treatment remain unknown. Here we show that the microglia-specific subtype of Ca(2+)-activated K(+) (BK) channel is responsible for generation of MIH and anti-nociceptive tolerance. We find that, after chronic morphine administration, an increase in arachidonic acid levels through the μ-opioid receptors leads to the sole activation of microglial BK channels in the spinal cord. Silencing BK channel auxiliary β3 subunit significantly attenuates the generation of MIH and anti-nociceptive tolerance, and increases neurotransmission after chronic morphine administration. Therefore, microglia-specific BK channels contribute to the generation of MIH and anti-nociceptive tolerance.
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor 555 Conjugate, clone A60, from mouse, ALEXA FLUOR 555