Merck
  • Nickel-NHC-catalyzed alpha-arylation of acyclic ketones and amination of haloarenes and unexpected preferential N-arylation of 4-aminopropiophenone.
Nickel-NHC-catalyzed alpha-arylation of acyclic ketones and amination of haloarenes and unexpected preferential N-arylation of 4-aminopropiophenone.
The Journal of organic chemistry (2007-06-15)
Kouki Matsubara, Keita Ueno, Yuji Koga, Kenji Hara
ABSTRACT
Arylation of both acyclic ketones and primary and secondary amines was achieved using a new, simple, stable, and easy-to-access nickel(II)-halide complex bearing mixed PPh3/N-heterocyclic carbene ligands as a catalyst precursor. Acyclic ketones were first arylated at the alpha-position with the nickel catalyst. On the other hand, less basic amines, such as diphenylamine and 4-aminobenzophenone, were more favorable in the catalytic amination of haloarenes than basic amines, contrary to previous reports. N-Arylation of 4-aminopropiophenone was found to proceed selectively without causing alpha-arylation of the ketone group.
Sigma-Aldrich
2,6-Diisopropylaniline, 97%