Merck
  • Neuroinflammation and structural injury of the fetal ovine brain following intra-amniotic Candida albicans exposure.
Neuroinflammation and structural injury of the fetal ovine brain following intra-amniotic Candida albicans exposure.
Journal of neuroinflammation (2016-02-05)
Daan R M G Ophelders, Ruth Gussenhoven, Martin Lammens, Benno Küsters, Matthew W Kemp, John P Newnham, Matthew S Payne, Suhas G Kallapur, Allan H Jobe, Luc J Zimmermann, Boris W Kramer, Tim G A M Wolfs
ABSTRACT
Intra-amniotic Candida albicans (C. Albicans) infection is associated with preterm birth and high morbidity and mortality rates. Survivors are prone to adverse neurodevelopmental outcomes. The mechanisms leading to these adverse neonatal brain outcomes remain largely unknown. To better understand the mechanisms underlying C. albicans-induced fetal brain injury, we studied immunological responses and structural changes of the fetal brain in a well-established translational ovine model of intra-amniotic C. albicans infection. In addition, we tested whether these potential adverse outcomes of the fetal brain were improved in utero by antifungal treatment with fluconazole. Pregnant ewes received an intra-amniotic injection of 10(7) colony-forming units C. albicans or saline (controls) at 3 or 5 days before preterm delivery at 0.8 of gestation (term ~ 150 days). Fetal intra-amniotic/intra-peritoneal injections of fluconazole or saline (controls) were administered 2 days after C. albicans exposure. Post mortem analyses for fungal burden, peripheral immune activation, neuroinflammation, and white matter/neuronal injury were performed to determine the effects of intra-amniotic C. albicans and fluconazole treatment. Intra-amniotic exposure to C. albicans caused a severe systemic inflammatory response, illustrated by a robust increase of plasma interleukin-6 concentrations. Cerebrospinal fluid cultures were positive for C. albicans in the majority of the 3-day C. albicans-exposed animals whereas no positive cultures were present in the 5-day C. albicans-exposed and fluconazole-treated animals. Although C. albicans was not detected in the brain parenchyma, a neuroinflammatory response in the hippocampus and white matter was seen which was characterized by increased microglial and astrocyte activation. These neuroinflammatory changes were accompanied by structural white matter injury. Intra-amniotic fluconazole reduced fetal mortality but did not attenuate neuroinflammation and white matter injury. Intra-amniotic C. albicans exposure provoked acute systemic and neuroinflammatory responses with concomitant white matter injury. Fluconazole treatment prevented systemic inflammation without attenuating cerebral inflammation and injury.
Sigma-Aldrich
Monoclonal Anti-CNPase antibody produced in mouse, clone 11-5B, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Interleukin-6 Antibody, clone 4B6, clone 4B6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Olig-2 Antibody, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, a.a. 82-87, culture supernatant, clone 12, Chemicon®
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Interleukin-6 Antibody, serum, Chemicon®
Pricing and availability is not currently available.