Merck
  • Cross-coupling of benzylic acetates with arylboronic acids: one-pot transformation of benzylic alcohols to diarylmethanes.
Cross-coupling of benzylic acetates with arylboronic acids: one-pot transformation of benzylic alcohols to diarylmethanes.
Chemical communications (Cambridge, England) (2005-12-01)
Ryoichi Kuwano, Masashi Yokogi
ABSTRACT
Benzylic acetates reacted with arylboronic acids in the presence of a DPEphos-[Pd(eta3-C3H5)Cl]2 catalyst when tert-amyl alcohol was used as a solvent, and the catalytic cross-couplings produced diarylmethanes in high yields (up to 94% isolated yield).