Merck
  • Relationship of Podoplanin and Glutathione S-transferases T1 Expression with Laryngeal Cancer.
Relationship of Podoplanin and Glutathione S-transferases T1 Expression with Laryngeal Cancer.
International journal of molecular and cellular medicine (2012-10-01)
Deniz Kanliada, Ender Coskunpinar, Kadir Serkan Orhan, Yasemin Musteri Oltulu, Mehmet Celik, Ayse Eren, Ilhan Yaylim, Kemal Deger
ABSTRACT
The aim of this study is to determine whether there is a role of podoplanin and glutathione S-transferases T1 (GST-T1) expression in laryngeal squamous cell carcinoma. The study was completed with 33 patients and gene expression analysis was performed by qRT-PCR. The podoplanin and GST-T1 expression patterns were analyzed to determine their correlation with clinicopathologic parameters of laryngeal cancer. Of all patients, 20 had supraglottic, and the remaining 13 had glottic laryngeal cancer. Increased expression of podoplanin was found in 14 tumor tissues, but GST-T1 expression was not detected. Podoplanin expression did not show any prediction for regional metastasis, thyroid cartilage invasion, lymphatic vessel invasion or tumor differentiation for laryngeal cancer, also there were no significant differences in podoplanin expression between glottic and supraglottic regions, but extracapsullar extension is almost statistically significant (p=0,05).
Product Description
Roche
High Pure RNA Tissue Kit, sufficient for 50 isolation(s), suitable for RT-PCR, suitable for Northern blotting
Roche
First Strand cDNA Synthesis Kit for RT-PCR (AMV), sufficient for 30 reactions (including 5 control reactions), kit of 1 (10 components), suitable for RT-PCR, hotstart: no, dNTPs included