Merck
  • Simplification of the Mitsunobu reaction. Di-p-chlorobenzyl azodicarboxylate: a new azodicarboxylate.
Simplification of the Mitsunobu reaction. Di-p-chlorobenzyl azodicarboxylate: a new azodicarboxylate.
Organic letters (2006-10-20)
Bruce H Lipshutz, David W Chung, Brian Rich, Ricardo Corral
ABSTRACT
Di-p-chlorobenzyl azodicarboxylate (DCAD) is introduced as a novel, stable, solid alternative to DEAD and DIAD for a variety of Mitsunobu couplings. DCAD/Ph(3)P-mediated reactions in CH(2)Cl(2) generate a readily separable hydrazine byproduct. [reaction: see text]
Sigma-Aldrich
Di-(4-chlorobenzyl)azodicarboxylate, 97%