Merck
  • Optimization of silylation using N-methyl-N-(trimethylsilyl)-trifluoroacetamide, N,O-bis-(trimethylsilyl)-trifluoroacetamide and N-(tert-butyldimethylsilyl)-N-methyltrifluoroacetamide for the determination of the estrogens estrone and 17alpha-ethinylestradiol by gas chromatography-mass spectrometry.
Optimization of silylation using N-methyl-N-(trimethylsilyl)-trifluoroacetamide, N,O-bis-(trimethylsilyl)-trifluoroacetamide and N-(tert-butyldimethylsilyl)-N-methyltrifluoroacetamide for the determination of the estrogens estrone and 17alpha-ethinylestradiol by gas chromatography-mass spectrometry.
Journal of chromatography. A (2006-02-01)
Ali Shareef, Michael J Angove, John D Wells
ABSTRACT
This paper reports an improved silylation procedure for simultaneous determination of the steroid hormones 17alpha-ethinylestradiol (EE2) and estrone (E1) using gas chromatography-mass spectrometry (GC-MS). This follows a re-assessment of some of the popular silylation procedures using N-methyl-N-trimethylsilyltrifluoroacetamide (MSTFA), N-O-bis-(trimethylsilyl)-trifluoroacetamide (BSTFA) and N-(tert-butyldimethylsilyl)-N-methyltrifluoroacetamide (MTBSTFA), which lead to the formation of trimethylsilyl (TMS) and tert-butyldimethylsilyl (TBS) derivatives. Silylation of EE2 using MSTFA or BSTFA+1% TMCS in ethyl acetate, acetonitrile and dichloromethane solvents produced multiple peaks corresponding to TMS-E1, and 3-mono-TMS-EE2 and/or 3,17-di-TMS-EE2 in variable proportions depending on the solvent used. When pyridine or dimethyl formamide solvents were used in the silylation of EE2 under the same reaction conditions, only 3,17-di-TMS-EE2 derivative was formed. Derivatization using MTBSTFA reagents using ethyl acetate, acetonitrile, dichloromethane, pyridine and dimethyl formamide resulted in almost 100% conversion of mono-TBS-EE2 to the TBS-E1. Therefore, typical methods used in some previous GC-MS determinations of E1 and EE2 in environmental water and/or sediment samples are subject to speculation. However, we can confirm that any of the TMS reagents can be used with either pyridine or dimethyl formamide under suitable reaction conditions.
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, >97%
Sigma-Aldrich
N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide with 1% tert-Butyldimethylchlorosilane, ≥95%
Supelco
MTBSTFA (with 1% t-BDMCS), ampule of 5 × 1 mL, (with 1% t-BDMCS), analytical standard, Cerilliant®
Supelco
MSTFA Reagent, ampule of 10 × 1.2 mL, analytical standard, Cerilliant®
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥98.5%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide with 1% trimethylchlorosilane, for GC derivatization, LiChropur
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, BioReagent, for silylations, LiChropur
Supelco
N,O-Bis(trimethylsilyl)trifluoroacetamide, for GC derivatization, LiChropur, ≥99.0%