Merck
  • An improved method to determine cell viability by simultaneous staining with fluorescein diacetate-propidium iodide.
An improved method to determine cell viability by simultaneous staining with fluorescein diacetate-propidium iodide.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society (1985-01-01)
K H Jones, J A Senft
ABSTRACT
A rapid, simultaneous double-staining procedure using fluorescein diacetate (FDA) and propidium iodide (PI) is described for use in the determination of cell viability in cell suspension. Air-dried slide preparations can be made from the cell suspensions so that an accurate estimate of the viability of the cells in the original suspension can be made up to 1 week later. Viable cells fluoresce bright green, while nonviable cells are bright red. Furthermore, when FDA-PI staining is compared to trypan blue dye exclusion as a method to determine cell viability, FDA-PI is found to be more consistent over prolonged periods of exposure to the dyes. Therefore, double staining with FDA-PI is a rapid, convenient, and reliable method to determine cell viability.
Sigma-Aldrich
Plant Cell Viability Assay Kit, Fluorescent staining system to highlight viable and non-viable cells