Merck
  • Proteomic analysis identified DJ-1 as a cisplatin resistant marker in non-small cell lung cancer.
Proteomic analysis identified DJ-1 as a cisplatin resistant marker in non-small cell lung cancer.
International journal of molecular sciences (2011-07-13)
Hua-Zong Zeng, Yi-Qing Qu, Wen-Jun Zhang, Bing Xiu, An-Mei Deng, Ai-Bin Liang
ABSTRACT
The aim of study is to identify cisplatin-resistance associated biomarkers for non-small cell lung cancers (NSCLC). We use two-dimensional electrophoresis (2-DE) combined with MALDI-TOF mass spectrometry to compare the proteome between lung cancer cell line A549 and its cisplatin-resistant subline A549/DDP. Nine cisplatin resistance-related proteins were identified, and DJ-1, one of the differently expressed proteins, was selected for further validation and evaluation. Immunohistochemical results demonstrated that high expression level of DJ-1 was associated with cisplatin resistance and a predictor for poor prognosis in 67 locally advanced NSCLC patients. Furthermore, in vitro results showed that silencing DJ-1 increased the proliferation inhibitory effect of cisplatin to A549/DDP cells. In conclusion, DJ-1 might play an important role in the resistibility to cisplatin, and it could also act as a novel candidate biomarker for predicting the response of NSCLC patients to cisplatin-based chemotherapy.
Sigma-Aldrich
MISSION® esiRNA, targeting human PARK7