Merck
  • Persistent overexpression of phosphoglycerate mutase, a glycolytic enzyme, modifies energy metabolism and reduces stress resistance of heart in mice.
Persistent overexpression of phosphoglycerate mutase, a glycolytic enzyme, modifies energy metabolism and reduces stress resistance of heart in mice.
PloS one (2013-08-21)
Junji Okuda, Shinnichiro Niizuma, Tetsuo Shioi, Takao Kato, Yasutaka Inuzuka, Tsuneaki Kawashima, Yodo Tamaki, Akira Kawamoto, Yohei Tanada, Yoshitaka Iwanaga, Michiko Narazaki, Tetsuya Matsuda, Souichi Adachi, Tomoyoshi Soga, Genzou Takemura, Hiroshi Kondoh, Toru Kita, Takeshi Kimura
ABSTRACT
Heart failure is associated with changes in cardiac energy metabolism. Glucose metabolism in particular is thought to be important in the pathogenesis of heart failure. We examined the effects of persistent overexpression of phosphoglycerate mutase 2 (Pgam2), a glycolytic enzyme, on cardiac energy metabolism and function. Transgenic mice constitutively overexpressing Pgam2 in a heart-specific manner were generated, and cardiac energy metabolism and function were analyzed. Cardiac function at rest was normal. The uptake of analogs of glucose or fatty acids and the phosphocreatine/Ī²ATP ratio at rest were normal. A comprehensive metabolomic analysis revealed an increase in the levels of a few metabolites immediately upstream and downstream of Pgam2 in the glycolytic pathway, whereas the levels of metabolites in the initial few steps of glycolysis and lactate remained unchanged. The levels of metabolites in the tricarboxylic acid (TCA) cycle were altered. The capacity for respiration by isolated mitochondria in vitro was decreased, and that for the generation of reactive oxygen species (ROS) in vitro was increased. Impaired cardiac function was observed in response to dobutamine. Mice developed systolic dysfunction upon pressure overload. Constitutive overexpression of Pgam2 modified energy metabolism and reduced stress resistance of heart in mice.
Sigma-Aldrich
L-Lactic Dehydrogenase from rabbit muscle, Type XI, lyophilized powder, 600-1,200 units/mg protein
Sigma-Aldrich
D-(āˆ’)-3-Phosphoglyceric acid disodium salt, ā‰„93% dry basis (enzymatic), powder
Sigma-Aldrich
Monoclonal ANTI-FLAGĀ® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)