Merck
Wrong gas/liquid partition data by gas chromatography.
Journal of chromatography. A (2006-02-25)
Ervin Sz Kováts, Richárd Kresz
ABSTRACT
In a paper published in 1992 [K.S. Reddy, J.-Cl. Dutoit, E.sz. Kováts, Pair-wise interactions by gas chromatography. I. Interaction free enthalpies of solutes with non-associated primary alcohol groups, J. Chromatogr. 609 (1992) 229] retention indices and standard chemical potential differences (SPOT-s) are given for about 160 solutes on a liquid branched paraffin hydrocarbon of the carbon number, z = 78. For the temperature dependence of the SPOT-s the proposal of Kirchhoff was accepted i.e. that at higher temperatures the molar heat capacity difference of the solute between the gas phase and the solution is nearly constant in a broad temperature range. The data were determined under conditions where the effect of adsorption could be neglected. By comparing the published data with those determined on another branched paraffin of the carbon number, z = 87 [F. Riedo, D. Fritz, G. Tarján, E.sz. Kováts, A tailor-made C87 hydrocarbon as a possible non-polar standard stationary phase for gas chromatography, J. Chromatogr. 126 (1976) 63] it is shown that the reported thermodynamic constants of solutes determined experimentally at higher temperatures (the group "H") are wrong on C78. In the present paper, the method of correction and the resulting corrected data are given. In later papers retention data are given on eight polar derivatives of the hydrocarbon C78 relative to those measured on the nonpolar standard [R. Cloux, G. Défayes, K. Fóti, J.-Cl. Dutoit, E.sz. Kováts, Pair-wise interactions by gas chromatography. III. Synthesis of isosteric stationary phases for gas chromatography, Synthesis (1993) 909; G. Défayes, K.S. Reddy, A. Dallos, E.sz. Kováts, Pair-wise interactions by gas chromatography. V. Interaction free enthalpies of solutes with primary chloro- and bromo-alkanes, J. Chromatogr. A 699 (1995) 131; K.S. Reddy, E.sz. Kováts, Pair-wise interactions by gas chromatography. II. Sociation free enthalpies of some hydrogen bonding solutes with non-associated primary alcohol groups, Chromatographia 34 (1992) 249; K.S. Reddy, R. Cloux, E.sz. Kováts, Pair-wise interactions by gas chromatography. IV. Interaction free enthalpies of solutes with trifluoromethyl-substituted alkanes, J. Chromatogr. A 673 (1994) 181; K.S. Reddy, R. Cloux, E.sz. Kováts, Pair-wise interactions by gas chromatography. VI. Interaction free enthalpies of solutes with primary methoxyalkane, cyanoalkane and alkanethiol groups, J. Chromatogr. A 704 (1995) 387; A. Dallos, A. Sisak, Z. Kulcsár, E.sz. Kováts, Pair-wise interactions by gas chromatography. VII. Interaction free enthalpies of solutes with secondary alcohol groups, J. Chromatogr. A 904 (2000) 211]. Obviously, the same data which are wrong in the nonpolar standard are also wrong in these solvents. The corrected data in the polar solvents are given in supplementary tables.
Sigma-Aldrich
1-Fluorooctane, 98%