Merck
  • Evolutions of volatile sulfur compounds of Cabernet Sauvignon wines during aging in different oak barrels.
Evolutions of volatile sulfur compounds of Cabernet Sauvignon wines during aging in different oak barrels.
Food chemistry (2016-02-28)
Dong-Qing Ye, Xiao-Tian Zheng, Xiao-Qing Xu, Yun-He Wang, Chang-Qing Duan, Yan-Lin Liu
ABSTRACT
The evolution of volatile sulfur compounds (VSCs) in Cabernet Sauvignon wines from seven regions of China during maturation in oak barrels was investigated. The barrels were made of different wood grains (fine and medium) and toasting levels (light and medium). Twelve VSCs were quantified by GC/FPD, with dimethyl sulfide (DMS) and methionol exceeding their sensory thresholds. Most VSCs tended to decline during the aging, while DMS was found to increase. After one year aging, the levels of DMS, 2-methyltetrahy-drothiophen-3-one and sulfur-containing esters were lower in the wines aged in oak barrels than in stainless steel tanks. The wood grain and toasting level of oak barrels significantly influenced the concentration of S-methyl thioacetate and 2-methyltetrahy-drothiophen-3-one. This study reported the evolution of VSCs in wines during oak barrel aging for the first time and evaluated the influence of barrel types, which would provide wine-makers with references in making proposals about wine aging.