Merck
  • Localization and expression of GABA transporters in the suprachiasmatic nucleus.
Localization and expression of GABA transporters in the suprachiasmatic nucleus.
The European journal of neuroscience (2015-09-24)
Michael Moldavan, Olga Cravetchi, Melissa Williams, Robert P Irwin, Sue A Aicher, Charles N Allen
ABSTRACT
GABA is a principal neurotransmitter in the suprachiasmatic hypothalamic nucleus (SCN), the master circadian clock. Despite the importance of GABA and GABA uptake for functioning of the circadian pacemaker, the localization and expression of GABA transporters (GATs) in the SCN has not been investigated. The present studies used Western blot analysis, immunohistochemistry and electron microscopy to demonstrate the presence of GABA transporter 1 (GAT1) and GAT3 in the SCN. By using light microscopy, GAT1 and GAT3 were co-localized throughout the SCN, but were not expressed in the perikarya of arginine vasopressin- or vasoactive intestinal peptide-immunoreactive (-ir) neurons of adult rats, nor in the neuronal processes labelled with the neurofilament heavy chain. Using electron microscopy, GAT1- and GAT3-ir was found in glial processes surrounding unlabelled neuronal perikarya, axons, dendrites, and enveloped symmetric and asymmetric axo-dendritic synapses. Glial fibrillary acidic protein-ir astrocytes grown in cell culture were immunopositive for GAT1 and GAT3 and both GATs could be observed in the same glial cell. These data demonstrate that synapses in the SCN function as 'tripartite' synapses consisting of presynaptic axon terminals, postsynaptic membranes and astrocytes that contain GABA transporters. This model suggests that astrocytes expressing both GATs may regulate the extracellular GABA, and thereby modulate the activity of neuronal networks in the SCN.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Anti-GABA Transporter-1 Antibody, CT (a.a. 588-599), Chemicon®, from rabbit
Sigma-Aldrich
Rabbit Anti-Mouse IgG Antibody, HRP conjugate, Chemicon®, from rabbit
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, Alexa Fluor488 conjugated, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein Antibody, clone GA5, clone GA5, Chemicon®, from mouse
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Atp11c
Sigma-Aldrich
Poly(ethyleneimine) solution, ~50% in H2O