Merck
  • Copper-mediated selective cyanation of indoles and 2-phenylpyridines with ammonium iodide and DMF.
Copper-mediated selective cyanation of indoles and 2-phenylpyridines with ammonium iodide and DMF.
Organic letters (2012-07-17)
Jinho Kim, Hyunwoo Kim, Sukbok Chang
ABSTRACT
Copper-mediated regioselective cyanation of indoles and 2-phenylpyridines was developed by using ammonium iodide and DMF as the combined source of a cyano unit under "Pd-free" conditions. Mechanistic studies indicate that the reaction of indoles proceeds through a two-step sequence: electrophilic initial iodination and then cyanation. The cyanation has a broad substrate scope, high functional group tolerance, and excellent regioselectivity.