Merck
  • Linear and cyclic peptides from the entomopathogenic bacterium Xenorhabdus nematophilus.
Linear and cyclic peptides from the entomopathogenic bacterium Xenorhabdus nematophilus.
Journal of natural products (2008-05-22)
Gerhard Lang, Tim Kalvelage, Arne Peters, Jutta Wiese, Johannes F Imhoff
ABSTRACT
Three new peptides, xenortides A and B and xenematide, were isolated from a culture of the nematode-associated entomopathogenic bacterium Xenorhabdus nematophilus. Their structures were elucidated using NMR, MS, and chemical derivatization methods. Xenortides A and B are the N-phenethylamide and tryptamide derivatives, respectively, of the dipeptide (NMe-L-Leu-NMe-L-Phe). The cyclodepsipeptide xenematide has the sequence (Thr-Trp-Trp-Gly), with a 2-phenylacetamide substituent at the threonine residue and one d-tryptophan. The new peptides and the two known compounds xenocoumacin II and nematophin were tested for antibacterial, antifungal, insecticidal, and anti-Artemia salina activities. Xenematide and xenocoumacin II showed moderate antibacterial activities. Xenocoumacin II, nematophin, and the two xenortides were active in the Artemia salina assay, and xenematide acted weakly insecticidal.
Product Description
Supelco
Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide, LiChropur, ≥98.0%