Sigma-Aldrich
2-Mercaptoethanol, BioUltra, for molecular biology, ≥99.0% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, random crystals, optical grade, 99.9% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Pricing and availability is not currently available.
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Pricing and availability is not currently available.
Sigma-Aldrich
MISSION® esiRNA, targeting human FKBP8
Pricing and availability is not currently available.
SAFC
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, for molecular biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, tablet
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, for molecular biology, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.85%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, BioUltra, for molecular biology, ~5 M in H2O
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 5 M
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, AnhydroBeads™, −10 mesh, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium chloride, BioUltra, for molecular biology, ≥99.5% (AT)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Pricing and availability is not currently available.
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri™, ≥99.5%
Pricing and availability is not currently available.
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Pricing and availability is not currently available.