Merck
  • Tumor Suppressive Function of p21-activated Kinase 6 in Hepatocellular Carcinoma.
Tumor Suppressive Function of p21-activated Kinase 6 in Hepatocellular Carcinoma.
The Journal of biological chemistry (2015-10-08)
Weisi Liu, Yidong Liu, Haiou Liu, Weijuan Zhang, Qiang Fu, Jiejie Xu, Jianxin Gu
ABSTRACT
Our previous studies identified the oncogenic role of p21-activated kinase 1 (PAK1) in hepatocellular carcinoma (HCC) and renal cell carcinoma (RCC). Contrarily, PAK6 was found to predict a favorable prognosis in RCC patients. Nevertheless, the ambiguous tumor suppressive function of PAK6 in hepatocarcinogenesis remains obscure. Herein, decreased PAK6 expression was found to be associated with tumor node metastasis stage progression and unfavorable overall survival in HCC patients. Additionally, overexpression and silence of PAK6 experiments showed that PAK6 inhibited xenografted tumor growth in vivo, and restricted cell proliferation, colony formation, migration, and invasion and promoted cell apoptosis and anoikis in vitro. Moreover, overexpression of kinase dead and nuclear localization signal deletion mutants of PAK6 experiments indicated the tumor suppressive function of PAK6 was partially dependent on its kinase activity and nuclear translocation. Furthermore, gain or loss of function in polycomb repressive complex 2 (PRC2) components, including EZH2, SUZ12, and EED, elucidated epigenetic control of H3K27me3-arbitrated PAK6 down-regulation in hepatoma cells. More importantly, negative correlation between PAK6 and EZH2 expression was observed in hepatoma tissues from HCC patients. These data identified the tumor suppressive role and potential underlying mechanism of PAK6 in hepatocarcinogenesis.
Sigma-Aldrich
MISSION® esiRNA, targeting human EZH2
Sigma-Aldrich
Nα-Acetyl-L-lysine
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Suz12
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Ezh2
Sigma-Aldrich
MISSION® esiRNA, targeting human SUZ12
Sigma-Aldrich
MISSION® esiRNA, targeting human EED
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Eed
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
Leptomycin B solution from Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)
Sigma-Aldrich
MISSION® esiRNA, targeting human PAK6, BUB1B-PAK6