Merck
TEMPO-Attached Pre-fluorescent Probes Based on Pyridinium Fluorophores.
Journal of fluorescence (2015-05-15)
Carolina Aliaga, Freddy Celis, Susan Lühr, Rubén Oñate
ABSTRACT
The photophysical behavior of three pyridinium-derived fluorophores, the N-aryl-2,4,6-triphenylpyridinium, the N-aryl-5,6-dihydro-2,4-diphenylbenzo[h]quinolinium and the N-aryl-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]acridinium perchlorates, was investigated. Comparison of their fluorescence quantum yields led to the preparation of a novel, more sensitive pyridinium-based, TEMPO-attached prefluorescent probe for H-abstraction processes, the N-{4-[4-(N-oxyl-2,2,6,6-tetramethylpiperidinyl)carbonylamino]phenyl}-5,6,8,9-tetrahydro-7-phenyldibenzo[c,h]-acridinium perchlorate.
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Benzaldehyde, purified by redistillation, ≥99.5%
Sigma-Aldrich
Benzaldehyde, ≥98%, FG, FCC
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Acetophenone, ≥98%, FG
Sigma-Aldrich
Benzaldehyde, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Acetophenone, natural, 98%, FG
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Acetophenone, ReagentPlus®, 99%
Sigma-Aldrich
Benzaldehyde, natural, FCC, FG
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis