Merck
  • Differential antitumor effects of vitamin D analogues on colorectal carcinoma in culture.
Differential antitumor effects of vitamin D analogues on colorectal carcinoma in culture.
International journal of oncology (2015-08-12)
J M Wierzbicka, A Binek, T Ahrends, J D Nowacka, A Szydłowska, Ł Turczyk, T Wąsiewicz, P M Wierzbicki, R Sądej, R C Tuckey, A T Slominski, J Chybicki, K Adrych, Z Kmieć, M A Żmijewski
ABSTRACT
Colorectal cancer (CRC) is an emerging global problem with the rapid increase in its incidence being associated with an unhealthy lifestyle. Epidemiological studies have shown that decreased levels of vitamin D3 significantly increases the risk of CRC. Furthermore, negative effects of vitamin D3 deficiency can be compensated by appropriate supplementation. Vitamin D3 was shown to inhibit growth and induce differentiation of cancer cells, however, excessive vitamin D3 intake leads to hypercalcemia. Thus, development of efficient vitamin D3 analogues with limited impact on calcium homeostasis is an important scientific and clinically relevant task. The aims of the present study were to compare the antiproliferative potential of classic vitamin D3 metabolites (1α,25(OH)2D3 and 25(OH)D3) with selected low calcemic analogues (calcipotriol and 20(OH)D3) on CRC cell lines and to investigate the expression of vitamin D-related genes in CRC cell lines and clinical samples. Vitamin D3 analogues exerted anti-proliferative effects on all CRC cell lines tested. Calcipotriol proved to be as potent as 1α,25(OH)2D3 and had more efficacy than 20-hydroxyvitamin D3. In addition, the analogs tested effectively inhibited the formation of colonies in Matrigel. The expression of genes involved in 1α,25(OH)2D3 signaling and metabolism varied in cell lines analysed, which explains in part their different sensitivities to the various analogues. In CRC biopsies, there was decreased VDR expression in tumor samples in comparison to the surgical margin and healthy colon samples (p<0.01). The present study indicates that vitamin D3 analogues which have low calcemic activity, such as calcipotriol or 20(OH)D3, are very promising candidates for CRC therapy. Moreover, expression profiling of vitamin D-related genes is likely to be a powerful tool in the planning of anticancer therapy. Decreased levels of VDR and increased CYP24A1 expression in clinical samples underline the importance of deregulation of vitamin D pathways in the development of CRC.
Sigma-Aldrich
25-Hydroxyvitamin D3 monohydrate, ≥99.0% (HPLC)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 50 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxyvitamin D3 solution, 5 μg/mL in ethanol, 98% (CP)
Sigma-Aldrich
25-Hydroxycholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
25-Hydroxyvitamin D3 solution, 100 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
1α,25-Dihydroxyvitamin D2 solution, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 50 μg/mL in ethanol, 95% (CP)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D2 solution, 100 μg/mL in ethanol, 95% (CP)