Merck
  • Thymic stromal lymphopoietin (TSLP) secretion from human nasal epithelium is a function of TSLP genotype.
Thymic stromal lymphopoietin (TSLP) secretion from human nasal epithelium is a function of TSLP genotype.
Mucosal immunology (2014-12-18)
C C K Hui, A Yu, D Heroux, L Akhabir, A J Sandford, H Neighbour, J A Denburg
ABSTRACT
Recent candidate gene and genome-wide association studies have identified "protective" associations between the single-nucleotide polymorphism (SNP) rs1837253 in the TSLP gene and risk for allergy, asthma, and airway hyperresponsiveness. The absence of linkage disequilibrium of rs1837253 with other SNPs in the region suggests it is likely a causal polymorphism for these associations, having functional consequences. We hypothesized that rs1837253 genotype would influence TSLP secretion from mucosal surfaces. We therefore evaluated the secretion of TSLP protein from primary nasal epithelial cells (NECs) of atopic and nonatopic individuals and its association with rs1837253 genotype. We found that although atopic sensitization does not affect the secretion of TSLP from NECs, there was decreased TSLP secretion in NECs obtained from heterozygous (CT; 1.8-fold) and homozygous minor allele (TT; 2.5-fold) individuals, as compared with NECs from homozygous major allele individuals (CC; P<0.05), after double-stranded RNA (dsRNA) stimulation (50 μg ml(-1)). Our novel results show that rs1837253 polymorphism may be directly involved in the regulation of TSLP secretion. This may help explain the protective association of this genetic variant with asthma and related traits. Identifying functional consequences of SNPs in genes with previously reported clinical associations is critical in understanding and targeting allergic inflammation.
Sigma-Aldrich
Trypan Blue, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
Trypan Blue, Dye content 60 %, ≥80% (HPLC)
Sigma-Aldrich
DL-Tryptophan, ≥99% (HPLC)
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
Trypan Blue solution, 0.4%, liquid, sterile-filtered, suitable for cell culture