Merck
  • Induction of apoptosis in prostate cancer cells by the novel ceramidase inhibitor ceranib-2.
Induction of apoptosis in prostate cancer cells by the novel ceramidase inhibitor ceranib-2.
In vitro cellular & developmental biology. Animal (2015-07-15)
Gokhan Kus, Selda Kabadere, Ruhi Uyar, Hatice Mehtap Kutlu
ABSTRACT
Ceramidases are key enzymes that decrease ceramide levels in cells. A reduction in ceramide concentration impairs ceramide signalling, and results in apoptosis resistance in cancer cells. This study investigates the potential for ceranib-2, a novel ceramidase inhibitor, to affect the survival and/or promote apoptosis of prostate cancer cells (LNCaP and DU145) in vitro. Cell viability was determined using MTT, and apoptosis assessed via flow cytometry. We examined structural changes with both confocal and transmission electron microscopes. Ceranib-2 concentrations of 0.1, 1, 5, 10, 25 and 50 μM were applied to LNCaP and DU145 cell lines. The corresponding reduction in LNCaP cell viability (against the control) was 84%, 80%, 64%, 56%, 40% and 15% after 24 h, and 81%, 74%, 60%, 55%, 27% and 11% after 48 h. For DU145 cells, viability was reduced to 84%, 82%, 63%, 50%, 41% and 18% after 24 h, and 64%, 42%, 30%, 20%, 8% and 5% after 48 h. Following treatment with 25 and 50 μM ceranib-2, the respective observed rates of early apoptosis in LNCaP cells were 23% and 36% after 24 h and 27% and 58% after 48 h. The morphological and ultrastructural signs of apoptosis detected were fragmented nuclei, chromatin condensations and cytoskeleton laceration. The inhibitory effects of ceranib-2 on prostate cancer cell survival are dose and time dependent. For LNCaP cells, ceranib-2 toxicity was predominately apoptotic in nature, while for DU145 cells, cell death may be related to non-apoptotic mechanisms.
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥98.0% (TLC)
Sigma-Aldrich
N-Acetyl-D-sphingosine, ≥97% (TLC), powder
Sigma-Aldrich
Fluorescein isothiocyanate isomer I, ≥97.5% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Fluorescein 5(6)-isothiocyanate, ≥90% (HPLC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%