Merck
  • Dehydroepiandrosterone prevents linoleic acid-induced endothelial cell senescence by increasing autophagy.
Dehydroepiandrosterone prevents linoleic acid-induced endothelial cell senescence by increasing autophagy.
Metabolism: clinical and experimental (2015-06-09)
Min Jung Lee, Eun Hee Kim, Sang Ah Lee, Yu Mi Kang, Chang Hee Jung, Hae Kyeong Yoon, So Mi Seol, Yoo La Lee, Woo Je Lee, Joong-Yeol Park
ABSTRACT
Autophagy has emerged as a potentially important factor in the pathogenesis of atherosclerosis. Dehydroepiandrosterone (DHEA) is an adrenal steroid of great recent interest due to its anti-aging and anti-atherogenic effects; however, little is known about its role in autophagy and endothelial senescence. The aim of this study was to investigate whether DHEA prevents linoleic acid (LA)-induced endothelial senescence by enhancing autophagy. After pre-treatement with or without DHEA prior to LA treatment in human aortic endothelial cells (HAECs), the level of senescence was compared by senescence-associated acidic β-galactosidase (SA-β-Gal) staining and hyperphosphorylated pRB (ppRB) protein level. Autophagy was detected by LC3 conversion and measuring the level of p62/SQSTM1 (sequestosome 1), a protein degraded by autophagy. The fusion of autophagosome and lysosome was confirmed by fluorescence microscopy. Pre-treatment with DHEA inhibited LA-induced endothelial senescence. DHEA increased the conversion of LC3-I to LC3-II and decreased the level of p62 in a time- and dose-dependent manner. Although both DHEA and LA treatment increased the conversion of LC3-I to LC3-II, treatment of LA increased p62 and decreased fusion of autophagosome and lysosome, which reflected decreased autophagic flux. However, pre-treatment with DHEA restored autophagic flux inhibited by LA. When we evaluated signaling pathways, we found that JNK activation involved in LC3 conversion induced by DHEA. DHEA prevents LA-induced endothelial senescence by restoring autophagy and autophagic flux through JNK activation.
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Anxa3
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Adenosine, suitable for cell culture, BioReagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Adenosine, ≥99%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Adenosine
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Linoleic acid, technical, 58-74% (GC)
Sigma-Aldrich
trans-Dehydroandrosterone, ≥99%
Sigma-Aldrich
Linoleic acid, ≥98%
Sigma-Aldrich
Linoleic Acid-Water Soluble, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
3-Methyladenine, autophagy inhibitor
Sigma-Aldrich
Linoleic acid, liquid, BioReagent, suitable for cell culture
Sigma-Aldrich
MISSION® esiRNA, targeting human ANXA3
Sigma-Aldrich
MISSION® esiRNA, targeting human MAP1LC3A
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)