Merck
  • Farnesoid X receptor knockdown provides significant growth inhibition in hepatocellular carcinoma cells while it does not interfere with the proliferation of primary human hepatocyte-derived cells.
Farnesoid X receptor knockdown provides significant growth inhibition in hepatocellular carcinoma cells while it does not interfere with the proliferation of primary human hepatocyte-derived cells.
The Journal of toxicological sciences (2015-07-15)
Tomofumi Fujino, Akiko Maruko-Ohtake, Yosuke Ohtake, Tomonori Kobayashi, Ken Ando, Airi Takeuchi, Yasuhito Ohkubo, Makio Hayakawa
ABSTRACT
Identification of substances with specific toxicity for carcinoma cells promises to facilitate the development of cancer chemotherapeutics that cause minimal side effects. Here, we show that knockdown of the farnesoid X receptor (FXR) effectively suppresses the proliferation of human hepatocellular carcinoma cell lines HepG2 and HLE accompanied by elevated expression of cyclin-dependent kinase (CDK) inhibitor p16/INK4a and p21/Cip1 proteins. On the other hand, the growth of the primary human hepatocyte-derived cell line Fa2N-4 is not affected by the treatment with FXR siRNA irrespective of marked increases in the mRNAs of p16/INK4a and p21/Cip1. Surprisingly, the expression levels of p16/INK and p21/Cip1 proteins are left unchanged in Fa2N-4 cells that are subjected to the FXR siRNA treatment. Since the expression levels of these CDK inhibitor proteins in FXR-knockdown Fa2N-4 cells were elevated in the presence of proteasomal inhibitor MG132, these CDK inhibitors may be subjected to the proteasomal degradation, thereby counteracting the increased expression of their cognate mRNAs, therefore similar levels of p16 and p21 proteins were observed in control and FXR-knockdown Fa2N-4 cells. These results suggest that FXR-knockdown is effective for inhibiting the proliferation of hepatocellular carcinoma cells, not interfering with the regulatory mechanism of normal hepatocyte growth.
Sigma-Aldrich
Thymidine, ≥99.0% (HPLC)
Sigma-Aldrich
GW4064, ≥97% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human NR1H4
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Dlc1
Sigma-Aldrich
MISSION® esiRNA, targeting human DLC1
Sigma-Aldrich
MISSION® esiRNA, targeting human HP
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin sulfate salt, powder
Sigma-Aldrich
Thymidine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thymidine, ≥99%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Nr1h4
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water