Merck
  • Far- and near-field properties of gold nanoshells studied by photoacoustic and surface-enhanced Raman spectroscopies.
Far- and near-field properties of gold nanoshells studied by photoacoustic and surface-enhanced Raman spectroscopies.
Physical chemistry chemical physics : PCCP (2015-01-07)
V Weber, A Feis, C Gellini, R Pilot, P R Salvi, R Signorini
ABSTRACT
Gold nanoshells, with a silica core and different core and shell dimensions, have been extensively investigated. Optical far-field properties, namely extinction and absorption, have been separately determined by means of spectrophotometry and photoacoustic spectroscopy, respectively, in the 440-900 nm range. The enhancement factor for surface-enhanced Raman scattering, which is related to near-field effects, has been measured from 568 to 920 nm. The absorption contribution to extinction decreases as the overall diameter increases. Moreover, absorption and scattering display different spectral distributions, the latter being red shifted. The Surface Enhanced Raman Scattering enhancement profile, measured using thiobenzoic acid as a Raman probe, is further shifted to the red. The latter result suggests that the enhancement is dominated by the presence of hot spots, which are possibly related to the surface roughness of gold nanoshell particles.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Thiobenzoic acid, technical grade, 90%
Sigma-Aldrich
Potassium carbonate, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Potassium carbonate, meets USP testing specifications
Sigma-Aldrich
Potassium carbonate, 99.995% trace metals basis
Sigma-Aldrich
Potassium carbonate, BioUltra, anhydrous, ≥99.0% (T)
Sigma-Aldrich
(3-Aminopropyl)trimethoxysilane, 97%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free