Merck
  • 5-Methoxytryptophan-dependent inhibition of oral squamous cell carcinoma metastasis.
5-Methoxytryptophan-dependent inhibition of oral squamous cell carcinoma metastasis.
Electrophoresis (2015-07-15)
Ssu-Han Wang, Chien-Wen Chang, Hsiu-Chuan Chou
ABSTRACT
The metastatic status of oral cancer is highly associated with the overall survival rate of patients. Previous studies have revealed that the endogenous tryptophan metabolite 5-methoxytryptophan (5-MTP) can downregulate cyclooxygenase-2 expression; suppress tumor proliferation, migration, and invasion; and reduce the tumor size. To improve the understanding of the molecular mechanisms involved in the regulation of 5-MTP in the tumorigenesis of oral cancer, we conducted a comparative wound healing and transwell invasion assays. Our results revealed that 5-MTP reduce oral cancer cell migration and invasion ability. In addition, the results of an in vivo assay demonstrated that the growth of primary tumors was significantly inhibited by 5-MTP in OC3 oral cancer cells and in invasive OC3-I5 oral cancer cells. Moreover, enlarged spleens were observed in OC3-I5-implanted severe combined immunodeficiency mice although 5-MTP can inhibit spleen enlargement. Through comparative proteomics, we identified 32 differentially regulated protein spots by using 2D-DIGE/MALDI-TOF MS analyses. Some of the differentially regulated proteins such as amadillo-repeat-containing X-linked protein 1, phosphoglycerate kinase 1, tropomyosin alpha-1, and tropomyosin alpha-4 may be associated with the 5-MTP-dependent inhibition of oral cancer growth and metastasis. We conclude that 5-MTP plays a crucial role in inhibiting in vitro and in vivo cancer invasion and metastasis.
Sigma-Aldrich
Amoxicillin, 95.0-102.0% anhydrous basis
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Tantalum(V) ethoxide, packaged for use in deposition systems