Merck
  • Phospholipase and Aspartyl Proteinase Activities of Candida Species Causing Vulvovaginal Candidiasis in Patients with Type 2 Diabetes Mellitus.
Phospholipase and Aspartyl Proteinase Activities of Candida Species Causing Vulvovaginal Candidiasis in Patients with Type 2 Diabetes Mellitus.
Journal of microbiology and biotechnology (2015-06-03)
Rasha H Bassyouni, Ahmed Ashraf Wegdan, Abdelsamie Abdelmoneim, Wessam Said, Fatma AboElnaga
ABSTRACT
Few research had investigated the secretion of phospholipase and aspartyl proteinase from Candida spp. causing infection in females with type 2 diabetes mellitus. This research aimed to investigate the prevalence of vulvovaginal candidiasis (VVC) in diabetic versus non-diabetic women and compare the ability of identified Candida isolates to secrete phospholipases and aspartyl proteinases with characterization of their genetic profile. The study included 80 females with type 2 diabetes mellitus and 100 non-diabetic females within the child-bearing period. Candida strains were isolated and identified by conventional microbiological methods and by API Candida. The isolates were screened for their extracellular phospholipase and proteinase activities by culturing them on egg yolk and bovine serum albumin media, respectively. Detection of aspartyl proteinase genes (SAP1 to SAP8) and phospholipase genes (PLB1, PLB2) were performed by multiplex polymerase chain reaction. Our results indicated that vaginal candidiasis was significantly higher among the diabetic group versus nondiabetic group (50% versus 20%, respectively) (p = 0.004). C. albicans was the most prevalent species followed by C. glabrata in both groups. No significant association between diabetes mellitus and phospholipase activities was detected (p = 0.262), whereas high significant proteinase activities exhibited by Candida isolated from diabetic females were found (82.5%) (p = 0.000). Non-significant associations between any of the tested proteinase or phospholipase genes and diabetes mellitus were detected (p > 0.05). In conclusion, it is noticed that the incidence of C. glabrata causing VVC is increased. The higher prevalence of vaginal candidiasis among diabetics could be related to the increased aspartyl proteinase production in this group of patients.
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
D-(+)-Glucose, analytical standard
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications