Merck
  • Lack of Contribution of Multidrug Resistance-associated Protein and Organic Anion-transporting Polypeptide to Pharmacokinetics of Regorafenib, a Novel Multi-Kinase Inhibitor, in Rats.
Lack of Contribution of Multidrug Resistance-associated Protein and Organic Anion-transporting Polypeptide to Pharmacokinetics of Regorafenib, a Novel Multi-Kinase Inhibitor, in Rats.
Anticancer research (2015-08-09)
Kazuo Hotta, Jun Ueyama, Yasuaki Tatsumi, Ikuto Tsukiyama, Yuka Sugiura, Hiroko Saito, Katsuhiko Matsuura, Takaaki Hasegawa
ABSTRACT
We investigated whether hepatic multidrug resistance-associated protein 2 (ABCC2) is involved in the hepatobiliary excretion of regorafenib, a novel multi-kinase inhibitor, using Sprague-Dawley (SD) rats and Eisai hyperbilirubinemic rats (EHBR) lacking the efflux transporter ABCC2. The involvement of organic anion-transporting polypeptide 1 (OATP1; OATP in humans) and OATP2 in the hepatic uptake of regorafenib and their protein levels in the liver were also investigated in the two rat groups. When regorafenib (5 mg/kg) was administered intravenously, the plasma concentrations of regorafenib were higher in EHBR than those in SD rats. However, the slope of the plasma concentration-time curves was the same for the two groups. Although the apparent biliary clearance of regorafenib in EHBR was lower than that of SD rats, no significant difference in the biliary excretion rate was observed between them, suggesting that regorafenib is not a substrate for ABCC2 and is not excreted into bile by ABCC2. It was also found that the contribution of biliary excretion to the systemic elimination of regorafenib is small. The protein-binding profiles of regorafenib were found to be linear in both rat groups. The binding potency, which was very high in both rat groups (>99.5%), was significantly higher in EHBR than that in SD rats. No significant differences in the plasma concentrations of unbound regorafenib were observed between the two rat groups, suggesting that the differences observed in the pharmacokinetic behaviors of regorafenib between the two rat groups were due to differences in protein-binding. When the protein levels of hepatic OATP1 and OATP2 were measured by immunoblot analysis, the expression of both transporters in EHBR was less than 40% of that in SD rats. The present results suggest that regorafenib is not a substrate for OATP1 and OATP2. These findings suggest the possibility that ABCC2-mediated hepatobiliary excretion and OATP1/OATP2-mediated hepatic uptake do not play important roles in the disposition of regorafenib.
Sigma-Aldrich
Methanol-12C, 99.95 atom % 12C
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Glycerol formal, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 10% in H2O
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Glycerol formal, 47-67% 5-hydroxy-1,3-dioxane basis (GC), 33-53% 4-hydroxymethyl-1,3-dioxolane basis (GC)
Sigma-Aldrich
Methanol solution, 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Methanol
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Ammonia, anhydrous, ≥99.98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Ammonia-14N, 99.99 atom % 14N
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, ≥99.0% (GC)