Merck
  • Mechanism of improved cellulosic bio-ethanol production from alfalfa stems via ambient-temperature acid pretreatment.
Mechanism of improved cellulosic bio-ethanol production from alfalfa stems via ambient-temperature acid pretreatment.
Bioresource technology (2015-07-06)
Shengfei Zhou, Troy M Runge
ABSTRACT
Model compounds and recalcitrant biomass were studied to elucidate the mechanism of ambient-temperature acid pretreatment of cellulosic biomass for bio-ethanol production. Pure cellulose, a pure hemicellulose and alfalfa stems were pretreated with sulfuric acid under ambient temperature with varied acid loading and time. Changes in water-soluble carbohydrates (WSCs) and chemical components of substrates were determined, and ethanol production via simultaneous saccharification and fermentation (SSF) was studied. The results showed significant amount of WSCs formed, and the WSCs increased with increasing acid loading and pretreatment time. The ethanol yields from pure cellulose were primarily affected by the added ash. Acid loading showed significant positive effect on ethanol production from alfalfa stems, whereas pretreatment time showed much weaker positive effect. However, non-significant amounts of WSCs were removed by washing of dried substrates. It was hypothesized to be because the WSCs adsorbed onto bulk substrates during the freeze-drying step, as supported by experimental results.
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Tetracycline, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium hydroxide, BioUltra, for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, for molecular biology, 10 M in H2O
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
3-Ethyl-2,4-pentanedione, mixture of tautomers, 98%
Sigma-Aldrich
Sodium hydroxide, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Nitrogen, ≥99.998%
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC), BioXtra
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)