Merck
  • TRPC1 regulates calcium-activated chloride channels in salivary gland cells.
TRPC1 regulates calcium-activated chloride channels in salivary gland cells.
Journal of cellular physiology (2015-04-23)
Yuyang Sun, Lutz Birnbaumer, Brij B Singh
ABSTRACT
Calcium-activated chloride channel (CaCC) plays an important role in modulating epithelial secretion. It has been suggested that in salivary tissues, sustained fluid secretion is dependent on Ca(2+) influx that activates ion channels such as CaCC to initiate Cl(-) efflux. However direct evidence as well as the molecular identity of the Ca(2+) channel responsible for activating CaCC in salivary tissues is not yet identified. Here we provide evidence that in human salivary cells, an outward rectifying Cl(-) current was activated by increasing [Ca(2+)]i, which was inhibited by the addition of pharmacological agents niflumic acid (NFA), an antagonist of CaCC, or T16Ainh-A01, a specific TMEM16a inhibitor. Addition of thapsigargin (Tg), that induces store-depletion and activates TRPC1-mediated Ca(2+) entry, potentiated the Cl(-) current, which was inhibited by the addition of a non-specific TRPC channel blocker SKF96365 or removal of external Ca(2+). Stimulation with Tg also increased plasma membrane expression of TMEM16a protein, which was also dependent on Ca(2+) entry. Importantly, in salivary cells, TRPC1 silencing, but not that of TRPC3, inhibited CaCC especially upon store depletion. Moreover, primary acinar cells isolated from submandibular gland also showed outward rectifying Cl(-) currents upon increasing [Ca(2+)]i. These Cl(-) currents were again potentiated with the addition of Tg, but inhibited in the presence of T16Ainh-A01. Finally, acinar cells isolated from the submandibular glands of TRPC1 knockout mice showed significant inhibition of the outward Cl(-) currents without decreasing TMEM16a expression. Together the data suggests that Ca(2+) entry via the TRPC1 channels is essential for the activation of CaCC.
Product Description
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
SAFC
BIS-TRIS
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Trpc3
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPC1
Sigma-Aldrich
MISSION® esiRNA, targeting human TRPC3
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Trpc1