Merck
  • Design, synthesis, and biological evaluation of benzoselenazole-stilbene hybrids as multi-target-directed anti-cancer agents.
Design, synthesis, and biological evaluation of benzoselenazole-stilbene hybrids as multi-target-directed anti-cancer agents.
European journal of medicinal chemistry (2015-03-31)
Jun Yan, Yueyan Guo, Yali Wang, Fei Mao, Ling Huang, Xingshu Li
ABSTRACT
To identify novel multi-target-directed drug candidates for the treatment of cancer, a series of benzoselenazole-stilbene hybrids were synthesised by combining the pharmacophores of resveratrol and ebselen. The biological assay indicated that all of the hybrids exhibited antiproliferative activities against four human cancer cell lines and demonstrated good TrxR inhibitory activities. The mechanism of cell apoptosis was investigated in G2/M cell cycle arrest induced by compound 6e and the apoptosis of the human liver carcinoma Bel-7402 cell line. The significant increase in intracellular ROS confirmed that compound 6e was capable of causing oxidative stress-induced apoptosis in cancer cells. Our results support the potential of compound 6e as a candidate for further studies examining the development of novel drugs for cancer treatment.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Potassium phosphate tribasic, reagent grade, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Thioredoxin Reductase from rat liver, buffered aqueous glycerol solution, ≥100 units/mg protein (Bradford)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis